Skip to Content
Merck
All Photos(2)

Key Documents

HPA021016

Sigma-Aldrich

Anti-PLA2G3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GIII sPLA2, Anti-Group 3 secretory phospholipase A2, Anti-Group III secretory phospholipase A2, Anti-Phosphatidylcholine 2-acylhydrolase GIII, Anti-sPLA2-III

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SPPEVTNMLWELLGTTCFKLAPPLDCVEGKNCSRDPRAIRVSARHLRRLQQRRHQLQDKGTDERQPWPSEPLRGPMSFYNQCLQLTQAARRPDRQQKSWSQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLA2G3(50487)

General description

The gene PLA2G3 (group 3 secretory phospholipase A2) is mapped to human chromosome 22q12.2. The protein localizes in the cytosolic punctuate structures.

Immunogen

Group 3 secretory phospholipase A2 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PLA2G3 (group 3 secretory phospholipase A2) is a secretory phospholipase. It is involved in the release of cellular arachidonate (AA). PLA2G3 is reported as a negative regulator of ciliogenesis. Absence of PLA2G3 results in cells showing centrosome-concentrated recycling endosomes. In human neuronal system, it participates in xanthine/xanthine oxidase (X/XOD)-mediated apoptotic cell death. Polymorphism in PLA2G3 is linked with Alzheimer′s disease (AD). The expression of PLA2G3 is deregulated in various malignant tissues.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74555

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela Re et al.
BMC bioinformatics, 7, 413-413 (2006-09-20)
Common fragile sites (cfs) are specific regions in the human genome that are particularly prone to genomic instability under conditions of replicative stress. Several investigations support the view that common fragile sites play a role in carcinogenesis. We discuss a
Vesna Brglez et al.
Biochimie, 107 Pt A, 114-123 (2014-10-07)
Secreted phospholipases A2 (sPLA2s) hydrolyse cell and lipoprotein phospholipid membranes to release free fatty acids and lysophospholipids, and can also bind to specific proteins. Several sPLA2s have been associated with various cancers, including prostate, colon, gastric, lung and breast cancers
Ana Martínez-García et al.
Journal of Alzheimer's disease : JAD, 22(4), 1181-1187 (2010-10-12)
Oxidative stress, which plays a critical role in the pathogenesis of neurodegenerative diseases such as Alzheimer's disease (AD), is intimately linked to aging, the best established risk factor for AD. Studies in neuronal cells subjected to oxidative stress, mimicking such
Joon Kim et al.
Nature, 464(7291), 1048-1051 (2010-04-16)
Primary cilia are evolutionarily conserved cellular organelles that organize diverse signalling pathways. Defects in the formation or function of primary cilia are associated with a spectrum of human diseases and developmental abnormalities. Genetic screens in model organisms have discovered core
Makoto Murakami et al.
The Journal of biological chemistry, 278(12), 10657-10667 (2003-01-11)
Here we report cellular arachidonate (AA) release and prostaglandin (PG) production by novel classes of secretory phospholipase A(2)s (sPLA(2)s), groups III and XII. Human group III sPLA(2) promoted spontaneous AA release, which was augmented by interleukin-1, in HEK293 transfectants. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service