Skip to Content
Merck
All Photos(2)

Key Documents

HPA015253

Sigma-Aldrich

Anti-SLC35A3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Golgi UDP-GlcNAc transporter, Anti-Solute carrier family 35 member A3, Anti-UDP-N-acetylglucosamine transporter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SSRSVLSPVVGTDAPDQHLELKKPQELKEMERLPLANEDKTMF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC35A3(23443)

General description

SLC35A3 (solute carrier family 35, member A3) is a nucleotide sugar transporter, and is also called UDP-N-acetylglucosamine transporter. It resides in the Golgi bodies, and has a ubiquitous expression pattern. This gene is localized to human chromosome 1p21, and codes for a hydrophobic protein which spans the membrane multiple times.

Immunogen

UDP-N-acetylglucosamine transporter recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SLC35A3 (solute carrier family 35, member A3) is responsible for the glycosylation of macromolecules. This protein, along with SLC35A2, is thought to transport UDP-Gal, in Golgi bodies, for glycosyltransferases. It is involved in the development of the bones of head and trunk, i.e. axial skeleton. Studies in cattle show that mutations in this gene are linked with complex vertebral malformation. Mutations in this gene are also linked with epilepsy, autism spectrum disorder and arthrogryposis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73106

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Ishida et al.
Journal of biochemistry, 126(1), 68-77 (1999-07-07)
We have cloned the human UDP-N-acetylglucosamine (UDP-GlcNAc) transporter cDNA, which was recognized through a homology search in the expressed sequence tags database (dbEST) based on its similarity to the human UDP-galactose transporter. The chromosomal location of the UDP-GlcNAc transporter gene
Simon Edvardson et al.
Journal of medical genetics, 50(11), 733-739 (2013-09-14)
The heritability of autism spectrum disorder is currently estimated at 55%. Identification of the molecular basis of patients with syndromic autism extends our understanding of the pathogenesis of autism in general. The objective of this study was to find the
Mariusz Olczak et al.
Biochemical and biophysical research communications, 434(3), 473-478 (2013-04-16)
The role of UDP-galactose transporter (UGT; SLC35A2) and UDP-N-acetylglucosamine transporter (NGT; SLC35A3) in glycosylation of macromolecules may be coupled and either of the transporters may partially replace the function played by its partner. The aim of this study was to
Bo Thomsen et al.
Genome research, 16(1), 97-105 (2005-12-14)
The extensive use of a limited number of elite bulls in cattle breeding can lead to rapid spread of recessively inherited disorders. A recent example is the globally distributed syndrome Complex Vertebral Malformation (CVM), which is characterized by misshapen and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service