Skip to Content
Merck
All Photos(1)

Documents

HEL 12469

94101201, human lung (fetal), Fibroblast

Synonym(s):

12469, HEL, HEL-12469, HEL12469, HEL1249

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

HEL 12469, from human lung(fetal), 94101201

biological source

human lung (fetal)

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Cell Line Origin

Human embryonic lung fibroblast

Cell Line Description

Fibroblasts fuse when infected with human CMV at a high input multiplicity.

DNA Profile

Not specified

Culture Medium

EMEM (EBSS) + 2 mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4, i.e., seeding at 1-2x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: All customers are required to complete a Cell line Release Authorisation form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service