Skip to Content
Merck
All Photos(1)

Documents

C6 Cell Line from rat

92090409, rat nerve (glial tumor), Neuronal

Synonym(s):

C-6 Cells, RGC-6 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

C6 Cell Line from rat, 92090409

biological source

rat nerve (glial tumor)

description

Rat glial tumor

growth mode

Adherent

karyotype

2n = 42

morphology

Neuronal

products

S-100 protein, somatotrophin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat glial tumour

Cell Line Description

Derived from a rat glial tumour induced by N-nitrosomethylurea. The cells produce S-100 protein which is found in the neural tissue of vertebrates and has been found in brain tumours. Synthesis of S-100 increases 10 fold as cells reach confluency.

Application

C6 cell line has been used to study the ability of 2-(2,4-dihydroxyphenyl)thieno-1,3-thiazin-4-one (BChTT) to inhibit cancer cell proliferation and identify mechanisms involved in its actions. It has also been used to study the cytotoxic and antioxidant properties of supercritical CO2 extract (SCE) of old man′s beard (Usnea barbata).
C6 cells are reported to produce elevated levels of glyceryl phosphate dehydrogenase activity in response to glucocorticoids

Culture Medium

Ham′s F12 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-3x100,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service