Skip to Content
Merck
All Photos(1)

Key Documents

GL-1

89072612, lizard lung, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

GL-1, 89072612

biological source

lizard lung

growth mode

Adherent

karyotype

Modal no. 37 and 38

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Lizard Gekko lung

Cell Line Description

From adult male Tokay Gekko (indigenous to Southeast Asia). Observed to undergo spontaneous chromosomal alterations from a normal diploid number of 38 to a bimodal distribution of chromosome number 38-40. Susceptible to a number of mammalian viruses at 36°C.

Application

Chromosome and virus studies

Culture Medium

EMEM (HBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4. seeding at 4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 30°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service