Skip to Content
Merck
All Photos(1)

Key Documents

G-401

87042204, human kidney, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

G-401, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 87042204

biological source

human kidney

packaging

tube of 5 μg 87042204-DNA-5UG
pkg of vial of cells 87042204-1VL

growth mode

Adherent

karyotype

2n = 46, diploid

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian rhabdoid tumour (formerly classified as Wilm′s tumour)

Cell Line Description

Derived from a tumour of a 3 month old male Caucasian. Highly transformed and grows in soft agar. Highly undifferentiated. G401 was originally described as a cell line derived from a Wilm?s tumour. Due to a change in the classification of such tumours, the cell line was examined by Garvin et al., 1993 and found to be more appropriately classified as derived from a rhabdoid tumour of the kidney.

Application

Transformation and tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,13
D13S317: 9,14
D16S539: 12
D5S818: 13
D7S820: 11,14
THO1: 8,9.3
TPOX: 8,11
vWA: 16

Culture Medium

McCoy′s 5a + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Change medium every 4 days.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service