MCC14/2 cell line human
10092303, human skin, Large, low contrast cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
Product Name
MCC14/2 cell line human, 10092303
biological source
human skin
description
Merkel carcinoma from human skin
growth mode
Not specified
karyotype
Not specified
morphology
Large, low contrast cells
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Merkel carcinoma
Cell Line Description
Merkel cell carcinoma cell line. Cell Bank Australia reference number: CBA1340.
Culture Medium
RPMI 1640 (with 2 mM L-Glutamine + 25 mM HEPES) + 15% FCS/FBS
Subculture Routine
Optimal split ratio 1:8 (seeding density 104 cells/cm2).Cells incubated at 37 °C with 5% CO2.Harvest the cells using 0.05% Trypsin/EDTA at 37 °C for 5 min.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Helen Leonard - Queensland Institute of Medical Research, Australia
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service