Skip to Content
Merck
All Photos(1)

Key Documents

55723

SAFC

1-(3-Isothiocyanatobenzyl)-4-[2-(3,4-dihydro-2H-1-benzopyran-6-yl)-5-oxazolyl]pyridinium bromide

BioReagent, suitable for fluorescence, ≥90% (HPCE)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20BrN3O2S
CAS Number:
Molecular Weight:
506.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

product line

BioReagent

Quality Level

Assay

≥90% (HPCE)

form

solid

fluorescence

λex 384 nm; λem 531 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[Br-].S=C=Nc1cccc(C[n+]2ccc(cc2)-c3cnc(o3)-c4ccc5OCCCc5c4)c1

InChI

1S/C25H20N3O2S.BrH/c31-17-27-22-5-1-3-18(13-22)16-28-10-8-19(9-11-28)24-15-26-25(30-24)21-6-7-23-20(14-21)4-2-12-29-23;/h1,3,5-11,13-15H,2,4,12,16H2;1H/q+1;/p-1

InChI key

ZLHULGGTEHEJGJ-UHFFFAOYSA-M

General description

Class of highly photostable, water soluble fluorescent labels with large Stoke′s shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient (Ε~>20,000 in EtOH).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Litak, P.T.;Kauffman, J.M.
Journal of Heterocyclic Chemistry, 31, 457-457 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service