Key Documents
Z802778
Hellma® fluorescence cuvettes, Micro
High Performance Quartz Glass, spectral range 200-2500 nm, pathlength 10x2 mm, chamber volume 400 μL
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
Pricing and availability is not currently available.
material
Suprasil® quartz
description
4 windows
manufacturer/tradename
Hellma 115F-QS
parameter
200-2500 nm spectral range
H × W × D
40 mm × 12.5 mm × 12.5 mm
chamber volume
400 μL
pathlength
10x2 mm
General description
with PTFE stopper
Packaging
Sold as packages containing a single cell (1EA = 1 cell)
Legal Information
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)