RPI-FA1-1
rFra a 1 (Fragaria ananassaallergen 1.0102), Strawberry
pkg of 250 μg
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
analytical standard
Assay
>95%
mol wt
Mw 19 kDa
packaging
pkg of 250 μg
manufacturer/tradename
InBio
format
neat
storage temp.
−20°C
General description
Source: Pichia pastoris
Formulation: 20mM Tris, 0.3M NaCl, pH 8.0. Sterile filtered.
Concentration: See product insert.
Purification: Multi-step chromatographic purification. Purity > 95 % by SDS-PAGE
Formulation: 20mM Tris, 0.3M NaCl, pH 8.0. Sterile filtered.
Concentration: See product insert.
Purification: Multi-step chromatographic purification. Purity > 95 % by SDS-PAGE
Disclaimer
Allergens are provided for research and commercial use in vitro: not for human in vivo or therapeutic use.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service