Skip to Content
Merck
All Photos(1)

Key Documents

ERMFA013

Charpy specimens (impact toughness)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®
certified by the BSC

manufacturer/tradename

JRC

application(s)

life science and biopharma

Looking for similar products? Visit Product Comparison Guide

Packaging

Set with 5 bars

Analysis Note

For more information please see:
ERM-FA013_cert
ERM-FA013_report

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service