Skip to Content
Merck
All Photos(1)

Key Documents

COND12800

Supelco

Conductivity Standard - 12800 μmhos/cm

Certified Reference Material

Synonym(s):

12800 μmhos/cm Conductivity Standard (500ml bottle)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

application(s)

environmental

format

single component solution

storage temp.

2-30°C

General description

This whole volume certified reference material (CRM) is produced and certified according to ISO 17034 and ISO/IEC 17025. The specific conductance value of 12800.0 ± 54 µmhos/ cm is determined using SI and higher order standards from NIST.

All information regarding the use of this CRM including traceability, homogeneity, uncertainty and safety is documented in the certificate of analysis (CoA).

This whole volume sample is delivered in 500 mL HDPE bottle.

Application

CRMfor conductivity meter calibration.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service