Skip to Content
Merck
All Photos(1)

Key Documents

41458

Sigma-Aldrich

Rebaudioside E

≥90.0% (HPLC)

Synonym(s):

(-)-Rebaudioside E, Kaur-16-en-18-oic acid, 13-[(2-O-β-D-glucopyranosyl-β-D-glucopyranosyl)oxy]-, 2-O-β-D-glucopyranosyl-β-D-glucopyranosyl ester, (4α)-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H70O23
CAS Number:
Molecular Weight:
967.01
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.28

Quality Level

Assay

≥90.0% (HPLC)

form

powder
solid

color

white to off-white

application(s)

food and beverages

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O[C@@H]8O[C@@H]([C@H]([C@@H]([C@H]8O)O)O)CO)O[C@]32C[C@]4([C@H]([C@]5([C@@H]([C@](CCC5)(C)C(=O)O[C@@H]6O[C@@H]([C@H]([C@@H]([C@H]6O[C@@H]7O[C@@H]([C@H]([C@@H]([C@H]7O)O)O)CO)O)O)CO)CC4)C)CC3)CC2=C

InChI

1S/C44H70O23/c1-17-11-43-9-5-22-41(2,7-4-8-42(22,3)40(59)66-38-34(30(55)26(51)20(14-47)62-38)64-36-32(57)28(53)24(49)18(12-45)60-36)23(43)6-10-44(17,16-43)67-39-35(31(56)27(52)21(15-48)63-39)65-37-33(58)29(54)25(50)19(13-46)61-37/h18-39,45-58H,1,4-16H2,2-3H3/t18-,19-,20-,21-,22+,23+,24-,25-,26-,27-,28+,29+,30+,31+,32-,33-,34-,35-,36+,37+,38+,39+,41-,42-,43-,44+/m1/s1

InChI key

RLLCWNUIHGPAJY-SFUUMPFESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rebaudioside E was isolated from Stevia rebaudiana leaves. Steviol glycosides are typically 200-350 times sweeter than sucrose, therefore Rebaudioside E (stevia sweetner) can potentially be used in the production of low and no-calorie products.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sidd Purkayastha et al.
Regulatory toxicology and pharmacology : RTP, 72(3), 646-657 (2015-05-25)
The hydrolysis of the steviol glycosides rebaudioside (Reb) A and E, as well as steviolbioside (a metabolic intermediate) to steviol was evaluated in vitro using human fecal homogenates from healthy Caucasian and Asian donors. Incubation of each of the Rebs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service