Skip to Content
Merck
All Photos(1)

Key Documents

AB2226

Sigma-Aldrich

Anti-MDGA1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

MAM domain-containing glycosylphosphatidylinositol anchor protein 1, GPI and MAM protein, GPIM, Glycosylphosphatidylinositol-MAM, MAM domain-containing protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
mouse, rat
technique(s):
western blot: suitable
citations:

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

species reactivity (predicted by homology)

human (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MDGA1(266727)

General description

Studies report that MDGA1 is a layer-specific marker and an area-specific marker, being expressed in layers 2/3 throughout the neocortex, but within the primary somatosensory area (S1), MDGA1 is also uniquely expressed in layers 4 and 6a. Comparisons with other markers, including cadherins, serotonin, cytochrome oxidase, RORß, and COUP-TF1, reveal unique features of patterned expression of MDGA1 within cortex and S1 barrels. Furthermore, during early stages of development, MDGA1 is expressed by Reelin- and Tbr1-positive Cajal–Retzius neurons that originate from multiple sources outside of neocortex and emigrate into it. At even earlier stages, MDGA1 is expressed by the earliest diencephalic and mesencephalic neurons, which appear to migrate from a MDGA1-positive domain of progenitors in the diencephalon and form a "preplate." These findings show that MDGA1 is a unique marker for studies of cortical lamination and area patterning and together with recent reports suggest that MDGA1 has critical functions in forebrain/midbrain development.

Specificity

This antibody recognizes the MAM domain of MDGA1.

Immunogen

Epitope: MAM domain
KLH-conjugated linear peptide corresponding to the MAM domain of MDGA1.

Application

Anti-MDGA1 Antibody detects level of MDGA1 & has been published & validated for use in Western Blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blot in untreated and PNGase F treated mouse P7 brain tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected MDGA1 in 5 µg of untreated and PNGase F treated mouse P7 brain tissue lysate. This antibody recognizes glycosolated (>160 kDa) (lane 1) and deglycosolated MDGA1 (lane 2).

Target description

~130kDa observed. UniProt describes 2 isoforms produced by alternative splicing at ~106 kDa (Isoform 1), ~107 kDa (Isoform 2). An uncharacterized band may be observed at ~48 kDa in some tissue lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Untreated and PNGase F treated mouse P7 brain tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service