Skip to Content
Merck
All Photos(1)

Key Documents

8.43654

Sigma-Aldrich

Phenyl acetic aldehyde

for synthesis

Synonym(s):

Phenyl acetic aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O
CAS Number:
Molecular Weight:
120.15
MDL number:
UNSPSC Code:
12352114
EC Index Number:
204-574-5
NACRES:
NA.22

vapor pressure

0.35 hPa ( 19 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

1550 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

mp

-10 °C

transition temp

flash point 68 °C (External MSDS)

solubility

slightly soluble 2.21 g/L

density

1.026-1.030 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

O=CCc1ccccc1

InChI

1S/C8H8O/c9-7-6-8-4-2-1-3-5-8/h1-5,7H,6H2

InChI key

DTUQWGWMVIHBKE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.026 - 1.030
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

154.4 °F - (External MSDS)

Flash Point(C)

68 °C - (External MSDS)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service