Skip to Content
Merck
All Photos(1)

Key Documents

124030

Sigma-Aldrich

Akt Inhibitor XIII, Isozyme-Selective, Akti2-1/2

The Akt Inhibitor XIII, Isozyme-Selective, Akti2-1/2 controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Akt Inhibitor XIII, Isozyme-Selective, Akti2-1/2, N-(2-(Diethylamino)ethyl)-3-(4-((4-(2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)-1-piperidinyl)methyl)phenyl)-2-phenyl-6-quinolinecarboxamide, 3HCl, N-(2-(Diethylamino)ethyl)-2-(4-((4-(2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)-1-piperidinyl)methyl)phenyl)-3-phe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H43N7O2 · 3HCl
Molecular Weight:
763.20
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (mixture of regioisomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 25 mg/mL
water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A highly cell-permeable and water soluble analog of Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2 (Cat. No. 124018) that acts as a plecktrin homology (PH)-domain dependent, non-ATP competitive and an allosteric inhibitor of Akt1/2 (IC50 = 560 nM, 390 nM and 7.8 µM in in vitro kinase assay; 526 nM, 730 nM and >10 µM in LNCaP cells for Akt1, Akt2 and Akt3, respectively) with ~ 80-fold greater selectivity over PKA, PKC and SGK (IC50 >50 µM). One of its potent regioisomers (C7-substituted) is shown to enhance induced caspase-3 activity in several tumor cells.
A highly cell-permeable and water soluble analog of Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2 (Cat. No. 124018) that acts as a plecktrin homology (PH)-domain dependent, non-ATP competitive and an allosteric inhibitor of Akt1/2 (IC50 = 560 nM, 390 nM and 7.8 µM in in vitro kinase assay; 526 nM, 730 nM and >10 µM in LNCaP cells for Akt1, Akt2 and Akt3, respectively) with ~ 80-fold greater selectivity over PKA, PKC and SGK (IC50 >50 µM). One of its potent regioisomers (C7-substituted) is shown to enhance induced caspase-3 activity in several tumor cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhao, Z., et al. 2008. Bioorg. Med. Chem. Lett.18, 49.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Naushad Akhtar et al.
PLoS pathogens, 16(12), e1009136-e1009136 (2020-12-29)
The level of CD40 expression on dendritic cells (DCs) plays a decisive role in disease protection during Leishmania donovani (LD) infection. However, current understanding of the molecular regulation of CD40 expression remains elusive. Using molecular, cellular and functional approaches, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service