Skip to Content
Merck
All Photos(1)

Key Documents

890830P

Avanti

16:0 DAP

Avanti Research - A Croda Brand 890830P, powder

Synonym(s):

1,2-dipalmitoyl-3-dimethylammonium-propane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H73NO4
CAS Number:
Molecular Weight:
595.98
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 200 mg (890830P-200mg)
pkg of 1 × 25 mg (890830P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890830P

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(CN(C)C)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C37H73NO4/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-36(39)41-34-35(33-38(3)4)42-37(40)32-30-28-26-24-22-20-18-16-14-12-10-8-6-2/h35H,5-34H2,1-4H3

Application

1,2-dipalmitoyl-3-dimethylammonium-propane (16:0 DAP) is suitable for the preparation of miRNA encapsulated liposomes.

Biochem/physiol Actions

1,2-dipalmitoyl-3-dimethylammonium-propane (16:0 DAP) is a cationic lipid and serves as a pH-sensitive transfection reagent.

Packaging

5 mL Amber Glass Screw Cap Vial (890830P-200mg)
5 mL Amber Glass Screw Cap Vial (890830P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moran Amit et al.
Nature, 578(7795), 449-454 (2020-02-14)
The solid tumour microenvironment includes nerve fibres that arise from the peripheral nervous system1,2. Recent work indicates that newly formed adrenergic nerve fibres promote tumour growth, but the origin of these nerves and the mechanism of their inception are unknown1,3.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service