Skip to Content
Merck
All Photos(2)

Key Documents

860650P

Avanti

C24:1 Ceramide (d17:1/24:1(15Z))

Avanti Research - A Croda Brand 860650P, powder

Synonym(s):

N-nervonoyl-D-erythro-sphingosine (C17 base)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H79NO3
CAS Number:
Molecular Weight:
634.07
UNSPSC Code:
12352100
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860650P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860650P

lipid type

sphingolipids
deuterated lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCC/C=C\CCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCC

InChI

1S/C41H79NO3/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-25-27-29-31-33-35-37-41(45)42-39(38-43)40(44)36-34-32-30-28-26-16-14-12-10-8-6-4-2/h17-18,34,36,39-40,43-44H,3-16,19-33,35,37-38H2,1-2H3,(H,42,45)/b18-17-,36-34+/t39-,40+/m0/s1

InChI key

YTXGSYSKVFVWFG-AHTJCSRDSA-N

General description

C24:1 Ceramide (d17:1/24:1(15Z)) is a synthetic ceramide derived from a C17 sphingosine with nervonic acid (a monounsaturated analog of lignoceric acid (24:0)).

Application

C24:1 Ceramide (d17:1/24:1(15Z)) has been used as an internal standard to measure ceramide content in the tissue samples by mass spectrometery.

Biochem/physiol Actions

Ceramides play an important role in cell signaling, cell differentiation, proliferation and apoptosis.

Packaging

5 mL Amber Glass Screw Cap Vial (860650P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnieszka U Blachnio-Zabielska et al.
International journal of molecular sciences, 19(12) (2018-12-14)
Ceramide accumulation in muscle and in liver is implicated in the induction of insulin resistance. Much less in known about the role of ceramide in adipose tissue. The aim of the present study was to elucidate the role of ceramide
Long-term follow-up of muscle lipid accumulation, mitochondrial activity and oxidative stress and their relationship with impaired glucose homeostasis in high fat high fructose diet-fed rats
Wang Y, et al.
The Journal of Nutritional Biochemistry, 64, 182-197 (2019)
Luisa Coderch et al.
American journal of clinical dermatology, 4(2), 107-129 (2003-01-30)
Ceramides are the major lipid constituent of lamellar sheets present in the intercellular spaces of the stratum corneum. These lamellar sheets are thought to provide the barrier property of the epidermis. It is generally accepted that the intercellular lipid domain
Ling Chen et al.
Oncology reports, 34(1), 447-454 (2015-05-16)
Endocannabinoids and ceramides have demonstrated growth inhibition, cell death induction and pro-apoptotic activity in cancer research. In the present study, we describe the profiles of two major endocannabinoids, ceramides, free fatty acids and relevant metabolic enzymes in 47 pairs of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service