Z743249
Julabo® bath spring clamps for Erlenmeyer flasks
fits 10 mL flask
All Photos(1)
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970601
General description
The individual spring clips can be positioned in the shaking water bath using the base tray for spring clips (Z743201-1EA). A maximum of 56 spring clips for 10 ml Erlenmeyer flasks can be mounted on a base tray.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.