Key Documents
Z104361
Tubing quick-disconnects
for tubing i.d., 3/8 ‑ 1/2in.
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
material
PE
feature
for tubing i.d. 3/8 ‑ 1/2in.
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art F19729-0000
General description
Consists of two serrated connectors which are joined securely through a male-female center taper. Connector of one size can be interchanged with other sizes to produce reducing combinations.
  • Easy-to-assemble barbed connectors require no tools
  • Ideal for use with flexible tubing
  • Handy on lines that require frequent disconnection