Key Documents
NORTCM250
Bruker tapered PTFE caps for Match tubes
for NORS255001
About This Item
UNSPSC Code:
41121800
NACRES:
SB.32
feature
for NORS255001
packaging
pack of 5 ea
manufacturer/tradename
Norell TCM250
General description
Bruker Match tube caps are specially machined from PTFE fluoropolymer, are color coded by tube size, and correspond to the color of the tube clamp in the Match Insert Assembly, This enables quick identification of matching components. The tube cap incorporates a slightly tapered inner diameter. Initially, the cap aligns and pushes easily onto the tube, it then becomes progressively tighter as the tube reaches full depth within the cap, creating a positive seal to preserve volatile sample solutions throughout short or long term storage.
Legal Information
Match is a trademark of Bruker Biospin Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)