All Photos(1)
About This Item
Linear Formula:
CH3CH(NH2)CH2CO2C2H5
CAS Number:
Molecular Weight:
131.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical grade
Assay
90%
form
liquid
refractive index
n20/D 1.4241 (lit.)
bp
60-61 °C/13 mmHg (lit.)
density
0.894 g/mL at 25 °C (lit.)
storage temp.
−20°C
SMILES string
CCOC(=O)CC(C)N
InChI
1S/C6H13NO2/c1-3-9-6(8)4-5(2)7/h5H,3-4,7H2,1-2H3
InChI key
HHIOOBJZIASBFF-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
107.6 °F - closed cup
Flash Point(C)
42 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service