Skip to Content
Merck
All Photos(2)

Key Documents

D56608

Sigma-Aldrich

3,4-Dichlorobenzaldehyde

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CHO
CAS Number:
Molecular Weight:
175.01
Beilstein:
636379
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

crystals

bp

247-248 °C (lit.)

mp

39-42 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccc(Cl)c(Cl)c1

InChI

1S/C7H4Cl2O/c8-6-2-1-5(4-10)3-7(6)9/h1-4H

InChI key

ZWUSBSHBFFPRNE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Chen et al.
Polymers, 11(6) (2019-06-07)
In this paper, we report chemical modifications of inulin by seven kinds of aromatic Schiff bases, which are different from their substituent groups. The obtained inulin derivatives were confirmed by FTIR, 1H NMR, and 13C NMR. Then, we studied their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service