Skip to Content
Merck
All Photos(1)

Key Documents

CDS000306

Sigma-Aldrich

6-Methyl-2-heptanone

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O
CAS Number:
Molecular Weight:
128.21
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

liquid

SMILES string

CC(C)CCCC(C)=O

InChI

1S/C8H16O/c1-7(2)5-4-6-8(3)9/h7H,4-6H2,1-3H3

InChI key

DPLGXGDPPMLJHN-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia Sarai Girón-Calva et al.
PloS one, 16(2), e0246696-e0246696 (2021-02-17)
Maize with enhanced β-carotene production was engineered to counteract pervasive vitamin A deficiency in developing countries. Second-generation biofortified crops are being developed with additional traits that confer pest resistance. These include crops that can produce Bacillus thuringiensis Berliner (Bt) insecticidal
Hye Won Kim et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-17)
Styrene can be formed by the microbial metabolism of bacteria and fungi. In our previous study, styrene was determined as a spoilage marker of Fuji apples decayed by Penicillium expansum, which is responsible for postharvest diseases. In the present study
De-Wei Chen et al.
Food chemistry, 286, 71-77 (2019-03-05)
Lipids, particularly phospholipids, are known to play a significant role in the characteristic aroma of the different meat species. Both neutral lipids and phospholipids were extracted from egg yolk and added to minced chicken (1% w/w) prior to cooking in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service