Skip to Content
Merck
All Photos(2)

Key Documents

900995

Sigma-Aldrich

1-Methyl-3-trifluoromethyl-1H-pyrazole-4-boronic acid pinacol ester

≥97%

Synonym(s):

1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)-1H-pyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16BF3N2O2
CAS Number:
Molecular Weight:
276.06
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Quality Level

Assay

≥97%

form

powder or crystals

mp

99-100 °C

SMILES string

FC(F)(F)C1=NN(C)C=C1B2OC(C)(C)C(C)(C)O2

InChI

1S/C11H16BF3N2O2/c1-9(2)10(3,4)19-12(18-9)7-6-17(5)16-8(7)11(13,14)15/h6H,1-5H3

InChI key

MUZZCKPFJNQCIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful boronic acid pinacol ester for synthesis of various bioactive small molecules. This product was previously listed as BML00234.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service