Skip to Content
Merck
All Photos(1)

Key Documents

731994

Sigma-Aldrich

N-Ethyl-d5-diethanolamine

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)2NC2D5
Molecular Weight:
138.22
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

97% (CP)

form

liquid

bp

246-252 °C (lit.)

density

1.051 g/mL at 25 °C

mass shift

M+5

SMILES string

[2H]C([2H])([2H])C([2H])([2H])N(CCO)CCO

InChI

1S/C6H15NO2/c1-2-7(3-5-8)4-6-9/h8-9H,2-6H2,1H3/i1D3,2D2

InChI key

AKNUHUCEWALCOI-ZBJDZAJPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

255.2 °F

Flash Point(C)

124 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service