Skip to Content
Merck
All Photos(2)

Key Documents

724149

Sigma-Aldrich

4-Chloro-2-nitrobenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl2NO4S
CAS Number:
Molecular Weight:
256.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

75-79 °C

functional group

chloro
nitro

SMILES string

[O-][N+](=O)c1cc(Cl)ccc1S(Cl)(=O)=O

InChI

1S/C6H3Cl2NO4S/c7-4-1-2-6(14(8,12)13)5(3-4)9(10)11/h1-3H

InChI key

LYESTQKHIPXVIK-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service