680966
2-Propylzinc bromide solution
0.5 M in THF
Synonym(s):
Isopropylzinc bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2CHZnBr
CAS Number:
Molecular Weight:
188.38
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
concentration
0.5 M in THF
density
0.963 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
CC(C)[Zn]Br
InChI
1S/C3H7.BrH.Zn/c1-3-2;;/h3H,1-2H3;1H;/q;;+1/p-1
InChI key
IKQCKJSWVAERRG-UHFFFAOYSA-M
Related Categories
Application
2-Propylzinc bromide solution (0.5 M in THF) has been used as a reagent in a protocol for the nickel-catalyzed dehalogenation of organic halides.
Legal Information
Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 3
Target Organs
Central nervous system, Respiratory system
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
-4.0 °F
Flash Point(C)
-20 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nickel-catalyzed hydrodehalogenation of aryl halides
Weidauer Maik, et al.
Journal of Organometallic Chemistry, 729, 53-59 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service