Skip to Content
Merck
All Photos(1)

Key Documents

593869

Sigma-Aldrich

PSS-Octa[(3-glycidyloxypropyl)dimethylsiloxy] substituted

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H136O36Si16
CAS Number:
Molecular Weight:
1931.11
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

refractive index

n20/D 1.459 (lit.)

Quality Level

solubility

H2O: insoluble
THF: soluble
chloroform: soluble
hexane: soluble
toluene: soluble

density

1.150 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(CCCOCC1CO1)O[Si]23O[Si]5(O[Si](C)(C)CCCOCC4CO4)O[Si]7(O[Si](C)(C)CCCOCC6CO6)O[Si](O[Si](C)(C)CCCOCC8CO8)(O2)O[Si]%10(O[Si](C)(C)CCCOCC9CO9)O[Si](O[Si](C)(C)CCCOCC%11CO%11)(O3)O[Si](O[Si](C)(C)CCCOCC%12CO%12)(O5)O[Si](O[Si](C)(C)CCCOCC%13CO%13)(O7)O%10

InChI

1S/C64H136O36Si16/c1-101(2,33-17-25-65-41-57-49-73-57)81-109-89-110(82-102(3,4)34-18-26-66-42-58-50-74-58)92-113(85-105(9,10)37-21-29-69-45-61-53-77-61)94-111(90-109,83-103(5,6)35-19-27-67-43-59-51-75-59)96-115(87-107(13,14)39-23-31-71-47-63-55-79-63)97-112(91-109,84-104(7,8)36-20-28-68-44-60-52-76-60)95-114(93-110,86-106(11,12)38-22-30-70-46-62-54-78-62)99-116(98-113,100-115)88-108(15,16)40-24-32-72-48-64-56-80-64/h57-64H,17-56H2,1-16H3

InChI key

HCMVSVWSKGFMKX-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.9 °F - closed cup

Flash Point(C)

74.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service