490008
Fmoc-Glu-OH-15N
98 atom % 15N
Synonym(s):
N-(9-Fluorenylmethoxycarbonyl)-L-glutamic-15N acid, L-Glutamic acid-15N, N-Fmoc
About This Item
Recommended Products
isotopic purity
98 atom % 15N
Quality Level
optical activity
[α]20/D −22°, c = 1 in DMF
technique(s)
bio NMR: suitable
mp
185-188 °C (lit.)
application(s)
peptide synthesis
functional group
Fmoc
mass shift
M+1
storage temp.
2-8°C
SMILES string
OC(=O)CC[C@H]([15NH]C(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O
InChI
1S/C20H19NO6/c22-18(23)10-9-17(19(24)25)21-20(26)27-11-16-14-7-3-1-5-12(14)13-6-2-4-8-15(13)16/h1-8,16-17H,9-11H2,(H,21,26)(H,22,23)(H,24,25)/t17-/m0/s1/i21+1
InChI key
QEPWHIXHJNNGLU-ZKBIZJTASA-N
Related Categories
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service