438111
Poly(propylene glycol) monobutyl ether
average Mn ~1,000
Synonym(s):
Butoxypolypropylene glycol, PPG-14 Butyl Ether, PPG-16 Butyl Ether, Poly(propylene oxide) monobutyl ether, Polyoxypropylene glycol butyl monoether, Polypropylene glycol butyl ether, Polypropylene glycol monobutyl ether
About This Item
Recommended Products
form
viscous liquid
mol wt
average Mn ~1,000
refractive index
n20/D 1.448
viscosity
140 cSt(20 °C)(lit.)
bp
>200 °C (lit.)
transition temp
pour point −40 °C
density
0.989 g/mL at 25 °C
SMILES string
CCCCO.CC(O)CO
InChI
1S/C10H22O3/c1-4-5-6-12-8-10(3)13-7-9(2)11/h9-11H,4-8H2,1-3H3
InChI key
CUVLMZNMSPJDON-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Features and Benefits
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
285.1 °F - closed cup
Flash Point(C)
140.6 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service