Skip to Content
Merck
All Photos(1)

Documents

42237

Sigma-Aldrich

Fmoc-His(Trt)-Wang resin

extent of labeling: ~0.7 mmol/g loading

Synonym(s):

Nα-Fmoc-N(im)-trityl-L-histidine 4-benzyloxybenzyl ester polymer-bound, PS-PHB-His(Trt)-Fmoc

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

Quality Level

extent of labeling

~0.7 mmol/g loading

particle size

80-160 μm

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service