Skip to Content
Merck
All Photos(1)

Key Documents

392936

Sigma-Aldrich

2,6-Dimethoxy-4-hydroxybenzaldehyde

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2(OCH3)2CHO
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

225 °C (dec.) (lit.)

functional group

aldehyde

SMILES string

COc1cc(O)cc(OC)c1C=O

InChI

1S/C9H10O4/c1-12-8-3-6(11)4-9(13-2)7(8)5-10/h3-5,11H,1-2H3

InChI key

HZWPJAZIRZFCGX-UHFFFAOYSA-N

General description

2,6-Dimethoxy-4-hydroxybenzaldehyde (4-hydroxy-2,6-dimethoxybenzaldehyde) is a p-hydroxybenzaldehyde derivative. Its synthesis by Vielsmeyer-Haack reaction and confirmation of the product formation by 1H NMR has been reported. Its structure has been investigated. The sodium salt of 4-hydroxy-2,6-dimethoxybenzaldehyde may be used to derivatize Merrifield resin to form resin-bound aldehyde.

Application

2,6-Dimethoxy-4-hydroxybenzaldehyde (4-hydroxy-2,6-dimethoxybenzaldehyde) may be used as a test compound to investigate the bactericidal activity of benzaldehydes against Campylobacter jejuni, Escherichia coli, Listeria monocytogenes and Salmonella enterica.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mendel Friedman et al.
Journal of food protection, 66(10), 1811-1821 (2003-10-24)
We evaluated the bactericidal activities of 35 benzaldehydes, 34 benzoic acids, and 1 benzoic acid methyl ester against Campylobacter jejuni, Escherichia coli O157:H7, Listeria monocytogenes, and Salmonella enterica when these compounds were substituted on the benzene ring with 0, 1
A Structure-Based Activation Model of Phenol-Receptor Protein Interactions.
Lee K.
Bull. Korean Chem. Soc., 18, 16-22 (1997)
Ming-Fu Cheng et al.
Journal of combinatorial chemistry, 6(1), 99-104 (2004-01-13)
A library of 1,4-benzodiazepine-2,5-dione dicarboxylate derivatives containing aryl substituents at N(1)- and N(4)-positions to mimic the amino acid residues of Try-13, Phe-14, and Asp-18 in endothelin-1 is established by using the starting materials of alpha-amino esters, hydroxybenzaldehydes, nitrobenzoyl chlorides, and
4-Hydroxy-2, 6-dimethoxybenzaldehyde.
Wuyts C, et al.
Acta Crystallographica Section E, Structure Reports Online, 61(1), 79-80 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service