Skip to Content
Merck
All Photos(1)

Key Documents

372374

Sigma-Aldrich

Mesitylene-d12

98 atom % D

Synonym(s):

1,3,5-Trimethylbenzene-d12

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D3(CD3)3
CAS Number:
Molecular Weight:
132.27
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.496 (lit.)

density

0.947 g/mL at 25 °C (lit.)

mass shift

M+12

SMILES string

[2H]c1c(c([2H])c(c([2H])c1C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6H,1-3H3/i1D3,2D3,3D3,4D,5D,6D

InChI key

AUHZEENZYGFFBQ-ZPMNDIOMSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyong Lu et al.
Molecules (Basel, Switzerland), 19(9), 14292-14303 (2014-09-13)
Gated molecular encapsulations, with baskets of type 1, are postulated to occur by the mechanism in which solvent molecule penetrates the inner space of 1, through one of its apertures, while the residing guest simultaneously departs the cavity. In the
F Aladar Bencsath et al.
Analytical and bioanalytical chemistry, 407(14), 4079-4090 (2015-03-23)
A headspace solid-phase microextraction gas chromatography-mass spectrometry (SPME GC-MS) method is described, to screen seafood for volatile organic compounds (VOCs) associated with petrochemical taint. VOCs are extracted from the headspace of heated sample homogenates by adsorption onto a SPME fiber

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service