Skip to Content
Merck
All Photos(2)

Key Documents

201901

Sigma-Aldrich

Chrysophenine

Dye content 65 %

Synonym(s):

Direct Yellow 12

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H26N4Na2O8S2
CAS Number:
Molecular Weight:
680.66
Colour Index Number:
24895
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 65%

λmax

389 nm

ε (extinction coefficient)

≥29000 at 389-401 nm in water at 0.02 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].CCOc1ccc(cc1)\N=N\c2ccc(\C=C\c3ccc(cc3S([O-])(=O)=O)\N=N\c4ccc(OCC)cc4)c(c2)S([O-])(=O)=O

InChI

1S/C30H28N4O8S2.2Na/c1-3-41-27-15-11-23(12-16-27)31-33-25-9-7-21(29(19-25)43(35,36)37)5-6-22-8-10-26(20-30(22)44(38,39)40)34-32-24-13-17-28(18-14-24)42-4-2;;/h5-20H,3-4H2,1-2H3,(H,35,36,37)(H,38,39,40);;/q;2*+1/p-2/b6-5+,33-31+,34-32+;;

InChI key

YQMJDPHTMKUEHG-RNVLWOMCSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chiang Hsiu-Mei et al.
Journal of hazardous materials, 161(2-3), 1384-1390 (2008-06-10)
Sludge adsorbent (SA) and commercial activated carbon fibers (ACFC and ACFT) were applied to Orange II and Chrysophenine (CH) adsorption (BET surface area: ACFC>ACFT>SA). ACFT was primarily in the micropore range, while SA was approximately 500 A (macropore) and 80

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service