Merck
  • [Synthesis and alpha-adrenolytic activities of 2-(1,4-benzodioxan-2-yl- methyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline derivatives].
[Synthesis and alpha-adrenolytic activities of 2-(1,4-benzodioxan-2-yl- methyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline derivatives].
Die Pharmazie (1990-06-01)
E Glusa, H Grüner, A Hagen, D Lohmann, H Foken
ABSTRACT
The synthesis of the title compounds starting from 2-Chlormethylbenzdioxan and Tetrahydroisochinolines is presented. Their actions on the platelet aggregation and the inhibition of alpha-adrenoceptors at the isolated rabbit aorta and the vas deferens of the guinea pig were investigated.
Sigma-Aldrich
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%