Skip to Content
Merck
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%

6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
Empirical Formula (Hill Notation):
C11H15NO2 · HCl
CAS Number:
Molecular Weight:
229.70

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing