1278109
USP
Fluorometholone acetate
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(6α, 11bβ)-17-(Acetyloxy)-9-fluoro-11-hydroxy-6-methylpregna-1,4-diene-3,20-dione, 6α-Methyl-9α-fluoro-17-acetoxy-21-deoxyprednisolone, 6α-Methyl-9α-fluoro-21-desoxyprednisolone-17-acetate, Fluorometholone 17-acetate, NSC 47438
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
fluorometholone
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
F[C@@]21[C@H]([C@H]4[C@@]([C@@](CC4)(OC(=O)C)C(=O)C)(C[C@@H]2O)C)C[C@@H](C3=CC(=O)C=C[C@@]31C)C
InChI
1S/C24H31FO5/c1-13-10-19-17-7-9-23(14(2)26,30-15(3)27)22(17,5)12-20(29)24(19,25)21(4)8-6-16(28)11-18(13)21/h6,8,11,13,17,19-20,29H,7,9-10,12H2,1-5H3/t13-,17-,19-,20-,21-,22-,23-,24-/m0/s1
InChI key
YRFXGQHBPBMFHW-SBTZIJSASA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Tobramycin and Fluorometholone Acetate Ophthalmic Suspension
Analysis Note
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service