SU860119
Crimp seals with PTFE/silicone septa
Ultra-clean, gold aluminum (with 5.5 mm center hole), diam. × thickness 11 mm × 1.3 mm, red PTFE/white silicone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
gold aluminum (with 5.5 mm center hole)
red PTFE/white silicone
feature
Ultra-clean
hardness
45 (Durometer: shore A)
packaging
pkg of 1000 ea
availability
not available in USA
diam. × thickness
11 mm × 1.3 mm
compatibility
for use with 11 mm crimp top
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service