Skip to Content
Merck
All Photos(1)

Documents

75932

Supelco

Isoxaflutole

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

5-Cyclopropyl-4-[2-(methylsulfonyl)-4-(trifluoromethyl)benzoyl]isoxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12F3NO4S
CAS Number:
Molecular Weight:
359.32
Beilstein:
8344543
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

−20°C

SMILES string

CS(=O)(=O)c1cc(ccc1C(=O)c2cnoc2C3CC3)C(F)(F)F

InChI

1S/C15H12F3NO4S/c1-24(21,22)12-6-9(15(16,17)18)4-5-10(12)13(20)11-7-19-23-14(11)8-2-3-8/h4-8H,2-3H2,1H3

InChI key

OYIKARCXOQLFHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Isoxaflutole is a member of the isoxazole herbicide family and is used for pre-emergence control of a wide range of important broadleaf and grass weeds in corn and sugarcane. This herbicide is an inhibitor of 4-hydroxyphenylpyruvate dioxygenase (HPPD) enzyme, which results in blockage of carotenoid biosynthesis, and thus causes bleaching in susceptible weeds. Isoxaflutole is characterized by low use rate, rapid dissipation, and very low mammalian toxicity.

According to Regulation (EC) No 1107/2009 (repealing Directive 91/414/EEC) isoxaflutole is approved for use in European Union (EU). As per Reg. (EU) 2015/845, the maximum residue level (MRL) for the presence of isoxaflutole (sum of isoxaflutole and its diketonitrile-metabolite, expressed as isoxaflutole) in various products of plant and animal origin is from 0.02 mg/kg – 0.2 mg/kg.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. Isoxaflutole CRM can also be used to: 
  • Determine the effects of temperature and moisture on dissipation, transformation, and mineralization in isoxaflutole controlled laboratory conditions
  • Evaluate the effect of organic matter, soil pH, Ca2+ concentration, and clay content on the sorption of isoxafutole and determine the sorptive behavior of isoxafutole by five different soils varying in physical and chemical properties
  • Investigate the effect of isoxaflutole on overall growth, nodulation, and nitrogen fixation capacity of one isoxaflutole tolerant and one isoxaflutole sensitive chickpea cultivar
  • Study the effect of soil properties on the degradation of isoxaflutole and the sorption−desorption of isoxaflutole and its diketonitrile degradate on soil
  • Evaluate the effect of a mixture of flufenacet and isoxaflutole on microorganism population, ecophysiological diversity index, colony development index, enzymatic activity of soil, and maize growth

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service