Skip to Content
Merck
All Photos(1)

Key Documents

59791

Supelco

Amino-functionalized silica gel spherical

75-200 μm particle size

Synonym(s):

Silica gel 110 NH2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
23201100
NACRES:
SB.52

grade

high-purity grade

form

powder

composition

carbon content, ≥3.0%
nitrogen content, ≥1.0%

technique(s)

LPLC: suitable

surface area

500 m2/g

loss

~5% loss on drying

matrix

Silica

matrix active group

amino-functionalized silica

particle size

75-200 μm

pore size

110 Å pore size

separation technique

hydrophilic interaction (HILIC)

Looking for similar products? Visit Product Comparison Guide

General description

Amino-functionalized silica gel spherical is monodisperse in size and shape. It is prepared and characterized by thermogravimetery, X-ray diffraction, FT-IR, scanning electron microscopy (SEM) and N2 adsorption-desorption techniques.

Application

Amino-functionalized silica gel spherical was used for the removal of heavy metal ions like Pb(II), Cd(II) and Ni (II) from a ternary aqueous solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>464.0 °F

Flash Point(C)

> 240 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of Ni(II), Cd(II), and Pb(II) from a ternary aqueous solution by amino functionalized mesoporous and nano mesoporous silica.
Heidaria A, Y. H.
Chemical Engineering Journal, 153, 1-3 (2009)
Synthesis, characterization and adsorption studies of several heavy metal ions on amino-functionalized silica nano hollow sphere and silica gel.
Najafia M, Y. Y.
Separation and Purification Technology, 85, 193-205 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service