Skip to Content
Merck
All Photos(2)

Documents

59683-U

Supelco

Supel BioSPME 96-Pin Device

pack of 10 ea

Synonym(s):

Supel BioSPME 96-Pin Device

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104018
NACRES:
NB.21

packaging

pack of 10 ea

Quality Level

technique(s)

solid phase microextraction (SPME): suitable

matrix active group

C18 coating

application(s)

clinical
pharmaceutical

compatibility

for use with automated robotic liquid handling systems

Looking for similar products? Visit Product Comparison Guide

General description

The Supel BioSPME 96-Pin Device combines BioSPME technology with high throughput free fraction determination for plasma protein binding studies and other free analyte analyses commonly performed by pharma and clinical laboratories. This fast, micro-sample prep device comprises a 96-pin universal format that fits into the wells of standard 96-well plates. The pins are coated with C18 particles and a patented binder that allows for small analytes of interest to bind, while larger macromolecules cannot. This extraction technique is shown to be more than three times faster than rapid equilibrium dialysis methods with improved data quality. Supel BioSPME 96-Pin Device workflow can be performed on automation robots via handling with standard gripper paddles, or alternatively can be achieved using a manual technique.

Analysis Note

small molecules having molecular weights up to 1 kDa and logP range from 1-5

Legal Information

Supel is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service