Skip to Content
Merck
All Photos(1)

Key Documents

48692

Supelco

Pentachlorophenol solution

certified reference material, 500 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
1285380
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

500 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-30°C

SMILES string

Oc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6HCl5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H

InChI key

IZUPBVBPLAPZRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satyendra Kumar Garg et al.
Environmental science and pollution research international, 20(4), 2288-2304 (2012-08-07)
It is the first report in which a novel psychrotrophic Pseudomonas putida SKG-1 strain was evaluated for simultaneous bioremediation of pentachlorophenol and Cr(6+) under various cultural and nutritional conditions. Pentachlorophenol (PCP) dechlorination products, bacterial structure, and functional groups were characterized
Paola Di Leo et al.
Journal of hazardous materials, 244-245, 303-310 (2012-12-29)
The existence of a lot of worldwide pentachlorophenol-contaminated sites has induced scientists to concentrate their effort in finding ways to degrade it. Therefore, an effective tool to decompose it from soil mixtures is needed. In this work the efficiency of
Jiajiang Lin et al.
Environmental pollution (Barking, Essex : 1987), 178, 433-440 (2013-05-01)
Pentachlorophenol (PCP) dissipation occurs naturally in flooded soils and although dissipation half-lives vary between soil profiles at the millimeter-scale the reason is poorly understood. Vertical variations of PCP dissipation were investigated in three typical Chinese paddy soils; Soil 1 (Umbraqualf)
Min Yang et al.
Journal of hazardous materials, 237-238, 63-70 (2012-09-12)
In this study, an imprinted silica matrix of pentachlorophenol (PCP) co-loaded with Fe(3)O(4) nanoparticles and ZnS:Mn(2+) quantum dots (QDs) was fabricated. The introduction of Fe(3)O(4) nanoparticles to the imprinted matrix provided an easy way to separate PCP under an external
Zhi-Guo Sheng et al.
Toxicology and applied pharmacology, 266(3), 335-344 (2012-11-28)
We have shown previously that exposing bacteria to wood preservatives pentachlorophenol (PCP) and copper-containing compounds together causes synergistic toxicity. However, it is not clear whether these findings also hold true in mammalian cells; and if so, what is the underlying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service