Skip to Content
Merck
All Photos(1)

Key Documents

47324

Supelco

1,2,4-Trimethylbenzene

analytical standard

Synonym(s):

Pseudocumene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CH3)3
CAS Number:
Molecular Weight:
120.19
Beilstein:
1903005
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

vapor density

4.1 (vs air)

vapor pressure

4.5 mmHg ( 37.7 °C)

CofA

current certificate can be downloaded

autoignition temp.

959 °F

expl. lim.

6.4 %

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.504 (lit.)

bp

168 °C (lit.)

mp

−44 °C (lit.)

density

0.876 g/mL at 20 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Cc1ccc(C)c(C)c1

InChI

1S/C9H12/c1-7-4-5-8(2)9(3)6-7/h4-6H,1-3H3

InChI key

GWHJZXXIDMPWGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sigma-Aldrich offers 1,2,4-trimethylbenzene for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analyses. Purity of the chemical is about 99% unless otherwise stated.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Richard H McKee et al.
International journal of toxicology, 29(3), 277-290 (2010-05-08)
This article reports the results of neurobehavioral tests on representative aromatic constituents, specifically C(9) to C(11) species. The testing evaluated effects in several domains including clinical effects, motor activity, functional observations, and visual discrimination performance. Exposures ranging from 600 to
Z Zheng et al.
Applied microbiology and biotechnology, 57(4), 572-578 (2002-01-05)
Laboratory batch experiments were performed with contaminated aquifer sediments and four soluble aromatic components of jet fuel to assess their biodegradation under anaerobic conditions. The biodegradation of four aromatic compounds, toluene, o-xylene, 1,2,4-trimethylbenzene (TMB), and naphthalene, separately or together, was
High-energy physics. Environmental report paralyzes Italian neutrino lab.
Alexander Hellemans et al.
Science (New York, N.Y.), 300(5626), 1634-1634 (2003-06-14)
D F Shakirov et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (4)(4), 13-16 (2003-03-18)
The authors' investigations show that red cells and tissues of animals exposed to a single 4-h inhalation of vapors of 1,2,4-trimethylbenzene in concentration 10 mg/m3 contained reduced amount of ATP, accumulated amounts of ADP, AMP in active inclusion of 32-P
D F Shakirov et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (1)(1), 26-28 (2003-03-26)
The paper presents the results of studies of the amounts of diene conjugates, malonic dialdehyde, and Schiff's bases, which are molecular LPO products in the tissues of the lung, liver, kidney in animals exposed to 4-hour inhalation of cyclic hydrocarbons

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service