Skip to Content
Merck
All Photos(1)

Key Documents

46855-U

Supelco

n-Hydrocarbon Mix

1000 μg/mL each component in dichloromethane: carbon disulfide (50:50), certified reference material

Synonym(s):

Florida n-Hydrocarbon Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL each component in dichloromethane: carbon disulfide (50:50)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
petroleum

format

multi-component solution

storage temp.

2-30°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Analyte

Description

    Decane
    Docosane
    Dodecane
    Dotriacontane
    Eicosane
    Hexacosane
    Hexadecane
    Hexatriacontane
    Octacosane
    Octadecane
    Octane
    Octatriacontane
    Tetracontane
    Tetracosane
    Tetradecane
    Tetratriacontane
    Triacontane
    See All (17)

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service