Skip to Content
Merck
All Photos(1)

Documents

22396

Supelco

High Capacity Gas Purifier, Replacement Converter Tube

1/8 in. fitting

Synonym(s):

CONVERTER TUBE W/RECHARGE 1/8 IN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

fitting

1/8 in. fitting

Looking for similar products? Visit Product Comparison Guide

General description

This replacement converter tube fits 110 V or 230 V clam-shell ovens. The converter tube must be at operating temperature (580 °C) inside a clam-shell oven for proper operation. Because the material inside is not active at room temperature, converter tube ends are not capped/plugged during shipment.

  • Length: 10 in. (25.4 cm)
  • Diameter: 1/2 in. (12.7 mm)
  • Max. Inlet Pressure: 150 psi
  • Max. Flow Rate: 1100 mL/min.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service