Skip to Content
Merck
All Photos(9)

Key Documents

ZRB2140

Sigma-Aldrich

Anti-TYK2 Antibody, clone 2H22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Non-receptor tyrosine-protein kinase TYK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2H22, recombinant monoclonal

description

2H22 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 133.65 kDa
observed mol wt ~135 kDa

purified by

using Protein A

species reactivity

human, monkey

species reactivity (predicted by homology)

porcine, bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... Tyk2(7297)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2H22 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Non-receptor tyrosine-protein kinase TYK2. It targets an epitope within 23 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the C-terminal half of human Non-receptor tyrosine-protein kinase TYK2.

Application

Quality Control Testing

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected TYK2 in Raji cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected TYK2 in Jurkat and COS-7 cell lysates.

Affinity Binding Assay: A representative lot of this antibody bound TYK2 peptide with a KD of 8.7 x 10-9 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected TYK2 in Human fallopian tube tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Non-receptor tyrosine-protein kinase TYK2 (UniProt: P29597; also known as Tyk2) is encoded by the Tyk2 gene (Gene ID: 7297) in human.TYK2 is a ubiquitously expressed member of the Janus kinase (JAK) family that is associated with receptors of type I interferon, interleukins 6, 10, 12, and 23 and plays a key role in the signal transduction of these cytokines. It is important in both innate and adaptive immune cells and is shown to be an essential component of maintaining normal immune responses. TYK2 is also involved in various other processes, including cell growth, development, and cell migration. It is reported to associate with growth factor receptors and activates STAT family of transcription factors. STAT proteins that homo- or heterodimerize following their phosphorylation translocate to the nucleus and regulate cytokine responsive genes. Its protein kinase 1 domain (pseudokinase domain), located in amino acids 589-875, mediates autoinhibition of the TYK2 kinase domain. Phosphorylation of tyrosine 1054 and 1055 induces kinase domain (aa 897-1176) activation. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Wu, P., et al. (2020). Front. Pediatr. 8; 253; Lupardus, PJ., et al. (2014). Proc. Natl. Acad. Sci. USA. 111 (22) 8025-8030; Yeh, TC., et al. (2000). Proc. Natl. Acad. Sci. USA. 97(16); 8991-8996; Rani, MR., et al. (1999). J. Biol. Chem. 274(45); 32507-32511).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service