Z290742
Vacuum filtration assembly replacement parts
#5 silicone stopper
All Photos(1)
About This Item
Recommended Products
material
silicone housing
feature
cap plug seal
packaging
pkg of 5 ea
neck diam.
27 mm
Compatibility
For 25 mm assemblies, 1-hole