Key Documents
TMO374320-BR
Nunc® Low Profile Universal Tubes in 2D bar coded latch rack
polypropylene, capacity (5.0 mL), flat bottom, external thread, untreated, case of 10 × 24 ea
Synonym(s):
barcoded tube, sample storage tube
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
material
flat bottom
polypropylene
packaging
case of 10 × 24 ea
manufacturer/tradename
Nunc 374320-BR
capacity
(5.0 mL)
Related Categories
General description
A robust solution for solid or liquid sample storage. Available in configurations that meet your tracking and packaging requirements, Thermo Scientific Nunc Low-Profile 5.0mL Externally-Threaded Universal Tubes feature 2D barcodes, graduations, a white patch for labeling, and a wide mouth design for easy addition and extraction of sample material. The externally-threaded design minimizes contamination risk in manual handling procedures. The tubes and rack are rated for storage down to vapor phase liquid nitrogen temperatures.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)