Skip to Content
Merck
All Photos(1)

Key Documents

SRP6265

Sigma-Aldrich

HMGB1/HMG1 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

HMG1, HMG3, HMGB1, SBP-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

0.22 μg/mL

mol wt

calculated mol wt 25.7 kDa
observed mol wt 28 kDa (DTT-reduced. Protein migrates due to glycosylation. Phe17 is the predicted N-terminus.)
observed mol wt 32 kDa

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HMGB1(3146)

General description

High-mobility group protein B1 (HMGB1), also known as high-mobility group protein 1 (HMG-1) and amphoterin, is a member of the HMGB family consisting of three members, HMGB1, HMGB2 and HMGB3. HMGB1 is a non-histone architectural chromosomal protein ubiquitously present in all vertebrate nuclei and binds double-stranded DNA without sequence specificity. However, it can shuttle between the nucleus and cytoplasm. The protein is secreted by activated monocytes and macrophages. HMGB1 is from the high mobility group protein super family. The gene is mapped to human chromosome 13q12.

Application

HMGB1 (high-mobility group protein B1) has been used to identify factors masking HMGB1 in human serum and plasma. It has been used to study HMGB1-mediated growth and motility in papillary thyroid cancer cells.
HMGB1/HMG1 human has been used as a positive control to analyse the expression of purified HMGB1 gene.

Biochem/physiol Actions

In the nucleus, HMGB1 (high-mobility group protein B1) works as a nucleosome stabilizer and transcriptional regulator. It also exhibits chaperone-like activity and suppresses aggregation of polyglutamine. It is also involved in inflammation and autophagy. HMGB1 can interacts with toll-like receptor (TLR) 2 and 4, interleukin-1β, chemokine (C-X-C motif) ligand 12 (CXCL12) and pathogen-associated molecular pattern (PAMP) molecules. It also works as a damage-associated molecular pattern molecule (DAMP). The mechanism of inflammation and damage is binding to TLR4, which mediates HMGB1-dependent activation of macrophage cytokine release. This positions HMGB1 at the intersection of sterile and infectious inflammatory responses. HMGB1 has been studied as a DNA vaccine adjuvant and a target for cancer therapy. In presence of Epstein-Barr virus (EBV) infection, HMGB1 is upregulated and is responsible for the proliferation of human nasopharyngeal carcinoma cells. It is involved in cancer cell migration and invasion.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HMGB1 induces the overexpression of miR-222 and miR-221 and increases growth and motility in papillary thyroid cancer cells.
Mardente S
Oncology Reports, 28, 2285-2289 (2012)
Molecular adjuvant HMGB1 enhances anti-influenza immunity during DNA vaccination.
Fagone P
Gene Therapy, 18, 1070-1077 (2011)
High mobility group box 1 (HMGB1) acts as an ?alarmin? to promote acute myeloid leukaemia progression.
Yasinska I M, et al.
Oncoimmunology, 7(6), e1438109-e1438109 (2018)
Factors masking HMGB1 in human serum and plasma.
Urbonaviciute V
Journal of Leukocyte Biology, 81, 67-74 (2007)
Fanny Henrioux et al.
International journal of molecular sciences, 24(13) (2023-07-14)
Dry eye inflammation is a key step in a vicious circle and needs to be better understood in order to break it. The goals of this work were to, first, characterize alarmins and cytokines released by ocular surface cells in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service